missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tris-(2-Carboxyethyl)phosphine, Hydrochloride (TCEP)

Product Code. 10286352
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10286352

Brand: Invitrogen™ T2556

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Tris-(2-Carboxyethyl)phosphine, Hydrochloride (TCEP)

Disufide crosslinks of cystines in proteins can be reduced to cysteine residues by TCEP (tris-(2-carboxyethyl)phosphine). Unlike DTT (dithiothreitol), TCEP does not contain thiols and therefore usually does not need to be removed prior to thiol modification.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Form Solid
Quantity 1 g
compliance-icons
Product Identifier
  • tris-(2-carboxyethyl)phosphine, hydrochloride (TCEP)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Tris(2-carboxyethyl)phosphonium chloride

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.