Learn More
Tris(dimethylamino)aluminum dimer, Thermo Scientific Chemicals
Preparation of aluminum nitrides
£132.00 - £471.00
Chemical Identifiers
CAS | 32093-39-3 |
---|---|
Molecular Formula | C12H36Al2N6 |
Molecular Weight (g/mol) | 318.425 |
MDL Number | MFCD00269811 |
InChI Key | JGZUJELGSMSOID-UHFFFAOYSA-N |
Synonym | tris dimethylamido aluminum iii, tris dimethylamino aluminum dimer, bis bis dimethylamino alumanyl dimethylamine, bis mu-dimethylamino tetrakis dimethylamino dialuminum |
PubChem CID | 16688802 |
IUPAC Name | N-[bis(dimethylamino)alumanyl]-N-methylmethanamine |
SMILES | CN(C)[Al](N(C)C)N(C)C.CN(C)[Al](N(C)C)N(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11329328
|
Thermo Scientific Alfa Aesar
043237.03 |
1 g |
£132.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11349328
|
Thermo Scientific Alfa Aesar
043237.06 |
5 g |
£471.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Preparation of aluminum nitrides
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
32093-39-3 | |
318.425 | |
JGZUJELGSMSOID-UHFFFAOYSA-N | |
16688802 | |
CN(C)[Al](N(C)C)N(C)C.CN(C)[Al](N(C)C)N(C)C |
C12H36Al2N6 | |
MFCD00269811 | |
tris dimethylamido aluminum iii, tris dimethylamino aluminum dimer, bis bis dimethylamino alumanyl dimethylamine, bis mu-dimethylamino tetrakis dimethylamino dialuminum | |
N-[bis(dimethylamino)alumanyl]-N-methylmethanamine |
Specifications
32093-39-3 | |
Brown to White | |
90°C (0.05 mmHg)(sublimation) | |
C12H36Al2N6 | |
MFCD00269811 | |
Moisture sensitive | |
JGZUJELGSMSOID-UHFFFAOYSA-N | |
N-[bis(dimethylamino)alumanyl]-N-methylmethanamine | |
16688802 | |
Powder |
87°C to 89°C | |
0.865 g/mL | |
21°C (70°F) | |
Al2[N(CH3)2]6 | |
UN3131 | |
tris dimethylamido aluminum iii, tris dimethylamino aluminum dimer, bis bis dimethylamino alumanyl dimethylamine, bis mu-dimethylamino tetrakis dimethylamino dialuminum | |
CN(C)[Al](N(C)C)N(C)C.CN(C)[Al](N(C)C)N(C)C | |
318.425 | |
318.42 | |
Tris (dimethylamino)aluminum dimer |
Safety and Handling
GHS H Statement
H228-H260-H314-H318
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P223-P231+P232-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
H228-H260-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
missing translation for 'storageNote' :
- Packaged under Argon
Recommended Storage : Ambient temperatures
- Tris(dimethylamino)aluminum dimer
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H280-Contains gas under pressure; may explode if heated.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only