missing translation for 'onlineSavingsMsg'
Learn More

TE Buffer, Tris-EDTA, 1X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™

Tris-EDTA (TE) is routinely used for suspending nucleic acid samples.

Brand:  Fisher BioReagents BP2476-100

19.90 GBP valid until 2024-12-31
Use promo code "21951" to get your promotional price.


 View more versions of this product

Product Code. 10021703

  • £23.35 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TE buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • 1X solution contains 10 mM tris and 1 mM EDTA
  • Filtered through a 5 micron filter and autoclaved
  • Tested for the absence of DNase, RNase, and protease
  • TE buffer is routinely used for resuspending nucleic acid samples
  • Intended for laboratory research use only
Emergency Overview: May cause eye, skin, and respiratory tract irritation. Avoid contact with skin and eyes. Do not breathe dust. Do not breathe vapors or spray mist. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Move to fresh air. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. If not breathing, give artificial respiration. Obtain medical attention.

NFPA:

  • Health:1
  • Flammability:0
  • Instability:0
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tris-EDTA
77-86-1, 60-00-4, 7647-01-0, 7732-18-5
7.4 to 8.1
100 mL
Filtered through a 5-micron filter.
Protease free
Molecular Biology
1X Solution, pH 7.4
Colorless
Liquid
DNase free
Poly Bottle
TE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers