missing translation for 'onlineSavingsMsg'
Learn More

TE Buffer, Tris-EDTA, 1X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™

Tris-EDTA (TE) is routinely used for suspending nucleic acid samples.

£24.80 - £72.10

Specifications

Chemical Name or Material Tris-EDTA
Name Note 1X Solution, pH 7.4
CAS 77-86-1, 60-00-4, 7647-01-0, 7732-18-5
Color Colorless
pH 7.4 to 8.1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10021703
View Documents
Fisher BioReagents
BP2476-100
100 mL Poly Bottle
£24.80
100mL
Please to purchase this item. Need a web account? Register with us today!
10031703
View Documents Promotion Details
Fisher BioReagents
BP2476-500
500 mL Poly Bottle
£72.10
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Save time and simplify your buffer preparation step by using ready-made Fisher BioReagents 1X TE buffer solution
  • Eliminates the hassle of dilution or waiting for powder to dissolve
  • 1X solution contains 10 mM tris and 1 mM EDTA
  • Filtered through a 5 micron filter and autoclaved
  • Tested for the absence of DNase, RNase, and protease
  • TE buffer is routinely used for resuspending nucleic acid samples
  • Intended for laboratory research use only
Emergency Overview: May cause eye, skin, and respiratory tract irritation. Avoid contact with skin and eyes. Do not breathe dust. Do not breathe vapors or spray mist. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Move to fresh air. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. If not breathing, give artificial respiration. Obtain medical attention.

NFPA:

  • Health:1
  • Flammability:0
  • Instability:0
Specifications

Specifications

Tris-EDTA
77-86-1, 60-00-4, 7647-01-0, 7732-18-5
7.4 to 8.1
DNase free
Protease free
Molecular Biology
1X Solution, pH 7.4
Colorless
Liquid
Filtered through a 5-micron filter.
TE
Videos
Safety and Handling

Safety and Handling

CAUTION!

May cause eye, skin, and respiratory tract irritation. Avoid contact with skin and eyes. Do not breathe dust. Do not breathe vapors or spray mist.

Recommended Storage : RT

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TE Buffer, Tris-EDTA, 1X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.