Learn More
Tris-Glycine, 10X Solution (Electrophoresis), Fisher BioReagents™
Brand:  Fisher BioReagents BP1306-4
185.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10356743
  • £222.58 / 4 L
Explore more special offers
Description
Description
Tris-Glycine is commonly used as a buffer in native polyacrylamide gel electrophoresis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tris-Glycine
Undesignated
Liquid
DNase free
DNase-, RNase- and Protease-Free
Protease free
Electrophoresis
10X Solution
77-86-1, 56-40-6
4 L
Filtered through a 0.2-micron filter.
Poly CUBE
TG
Components: Tris (2-3%) and Glycine (13-14%).
0.25M Tris Base and 1.92M Glycine.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Tris-Glycine 10X Solution