missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris-Glycine, 10X Solution (Electrophoresis), Fisher BioReagents™
£71.20 - £196.00
Specifications
Chemical Name or Material | Tris-Glycine |
---|---|
Name Note | 10X Solution |
Color | Undesignated |
CAS | 77-86-1, 56-40-6 |
Physical Form | Liquid |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10746834
|
Fisher BioReagents
BP1306-1 |
1 L | Nalgene Poly Bottle |
£71.20
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10356743
|
Fisher BioReagents
BP1306-4 |
4 L | Poly CUBE | ||||||
Description
Tris-Glycine is commonly used as a buffer in native polyacrylamide gel electrophoresis.Specifications
Tris-Glycine | |
Undesignated | |
Liquid | |
Filtered through a 0.2-micron filter. | |
Protease free | |
Electrophoresis |
10X Solution | |
77-86-1, 56-40-6 | |
DNase free | |
DNase-, RNase- and Protease-Free | |
TG | |
Components: Tris (2-3%) and Glycine (13-14%). 0.25M Tris Base and 1.92M Glycine. |
Safety and Handling
Recommended Storage : RT