Learn More
Tris-Glycine, 10X Solution (Electrophoresis), Fisher BioReagents™
Brand:  Fisher BioReagents BP1306-1
68.36 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
 View more versions of this product
Product Code. 10746834
  • £75.60 / 1 L
Explore more special offers
Description
Description
Tris-Glycine is commonly used as a buffer in native polyacrylamide gel electrophoresis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tris-Glycine
Undesignated
Liquid
DNase free
DNase-, RNase- and Protease-Free
Protease free
Electrophoresis
10X Solution
77-86-1, 56-40-6
1 L
Filtered through a 0.2-micron filter.
Nalgene Poly Bottle
TG
Components: Tris (2-3%) and Glycine (13-14%).
0.25M Tris Base and 1.92M Glycine.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Tris-Glycine 10X Solution
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Tris-Glycine, 10X Solution (Electrophoresis), Fisher BioReagents™ > Nalgene Poly Bottle; 1L