missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Hydrochloride, 1M Solution (pH 7.0/Mol. Biol.), Fisher BioReagents™
£42.30 - £82.35
Chemical Identifiers
CAS | 77-86-1, 1185-53-1, 7732-18-5 |
---|---|
Molecular Formula | C4H11NO3 |
Molecular Weight (g/mol) | 121.136 |
InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
Synonym | Tris-HCl |
PubChem CID | 6503 |
ChEBI | CHEBI:9754 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol |
SMILES | C(C(CO)(CO)N)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10274773
|
Fisher BioReagents
BP1756-100 |
100 mL | Poly Bottle |
£42.30
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10543145
|
Fisher BioReagents
FLBP1756-500 |
500 mL | Poly Bottle |
£82.35
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Tris Hydrochloride, 1M Solution (pH 7.0/Mol. Biol.)
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. These 1M solutions of Tris Hydrochloride at pH 7.0 provide convenient and highly purified solutions.Chemical Identifiers
77-86-1, 1185-53-1, 7732-18-5 | |
121.136 | |
Tris-HCl | |
CHEBI:9754 | |
C(C(CO)(CO)N)O |
C4H11NO3 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
6503 | |
2-amino-2-(hydroxymethyl)propane-1,3-diol |
Specifications
77-86-1, 1185-53-1, 7732-18-5 | |
0.96 to 1.04 M | |
Liquid | |
Filtered through a 0.2-micron filter. | |
C4H11NO3 | |
Tris-HCl | |
C(C(CO)(CO)N)O | |
121.136 | |
CHEBI:9754 | |
Tris Hydrochloride, 1M Solution, pH 7.0 |
Undesignated | |
10 | |
DNase free | |
Pass Test | |
Protease free | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol | |
6503 | |
DNase-, RNase- and Protease-Free |
Safety and Handling
Recommended Storage : RT