missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Hydrochloride, 1M Solution (pH 7.0/Mol. Biol.), Fisher BioReagents™
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
Description
Tris Hydrochloride, 1M Solution (pH 7.0/Mol. Biol.)
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. These 1M solutions of Tris Hydrochloride at pH 7.0 provide convenient and highly purified solutions.

Specifications
Specifications
Color | Undesignated |
Concentration | 0.96 to 1.04 M |
pH | 10 |
Physical Form | Liquid |
Quantity | 100 mL |
DNase | DNase free |
Filtered Through | Filtered through a 0.2-micron filter. |
Identification | Pass Test |
Packaging | Poly Bottle |
Protease | Protease free |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
20Minutes
52Seconds
6 -
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
0Hours
20Minutes
52Seconds
6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tris Hydrochloride, 1M Solution (pH 7.0/Mol. Biol.), Fisher BioReagents™ >
Spot an opportunity for improvement?Share a Content Correction