missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Triton X-100 (1%)

Triton X-100 (1%)

Brand:  Invitrogen™ HFH10


Product Code. 10717503

  • £71.25 / Each
In Stock
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

This reagent, a component of the Fetal Hemoglobin (HbF) test, is a 0.1% working solution prepared in phosphate-buffered saline (PBS) containing bovine serum albumin (BSA) and sodium azide.

Class: IVD
Statement: For In Vitro Diagnostic Use.
PContains: 0.1% sodium azide
Stabilizer: 0.1% BSA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Detergent
Wet Ice
Triton X-100
Store in refrigerator (2–8°C).

Shelf life: 24 months from date of manufacture
Liquid
10 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1% Triton X-100

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : .alpha.-[4-(1,1,3,3-tetramethylbutyl)phenyl]-.omega.-hydroxy-Poly(oxy-1,2-ethanediyl)

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.