missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Triton™ X-100 Surfact-Amps™ Detergent Solution

Thermo Scientific™ Surfact-Amps X-100 Detergent Solution is highly-purified Triton X-100 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.

Brand:  Thermo Scientific™ 28314


 View more versions of this product

Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11891445

  • £147.00 / Pack of 6
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Triton X-100 Detergent Surfact-Amps Solution is highly-purified Triton X-100 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.

This is an easy-to-use 10% (w/v) solution of purified Triton™ X-100 detergent for use in routine and high-demand protein research methods and molecular biology techniques. Triton X-100 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins.

Surfact-Amps™ Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Highlights:

  • Triton X-100 – a nonionic detergent for use in various protein methods

  • Accurate – precise 10% detergent solution in ultrapure water

  • Easy-to-use – solution is simple to dispense and dilute for use

  • Exceptionally pure – less than 1.0μeq/ml peroxides and carbonyls

  • Stable – packed in glass ampules under inert nitrogen gas

Detergent Properties:

  • Molecular Weight: 647
  • Detergent Class: Nonionic
  • Aggregation Number: 140
  • Micelle Molecular Weight: 90,000
  • Critical Micelle Concentration (CMC): 0.24mM (0.0155%, w/v)
  • Cloud Point: 64°C
  • Dialyzable: No

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Detergent
Liquid
Detergent Solution
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.
Surfact-Amps™, Triton™
6 x 10 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Triton X-100 Surfact-Amps Detergent Solution

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Poly(oxy-1,2-ethanediyl),

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.