Learn More
Thermo Scientific™ Triton™ X-100 Surfact-Amps™ Detergent Solution
Thermo Scientific™ Surfact-Amps X-100 Detergent Solution is highly-purified Triton X-100 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.
Brand: Thermo Scientific™ 85111
100.00 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.
View more versions of this product
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Thermo Scientific™ Triton X-100 Detergent Surfact-Amps Solution is highly-purified Triton X-100 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.
This is an easy-to-use 10% (w/v) solution of purified Triton™ X-100 detergent for use in routine and high-demand protein research methods and molecular biology techniques. Triton X-100 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins.
Surfact-Amps™ Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.
Highlights:
Triton X-100 – a nonionic detergent for use in various protein methods
Accurate – precise 10% detergent solution in ultrapure water
Easy-to-use – solution is simple to dispense and dilute for use
Exceptionally pure – less than 1.0μeq/ml peroxides and carbonyls
Stable – packed in glass ampules under inert nitrogen gas
Detergent Properties:
- Molecular Weight: 647
- Detergent Class: Nonionic
- Aggregation Number: 140
- Micelle Molecular Weight: 90,000
- Critical Micelle Concentration (CMC): 0.24mM (0.0155%, w/v)
- Cloud Point: 64°C
- Dialyzable: No

Specifications
Detergent | |
Liquid | |
Detergent Solution |
Store between 20°C and 25°C | |
Surfact-Amps™, Triton™ | |
50 mL |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Triton X-100 Surfact-Amps Detergent Solution, 50 mL.
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
Hazard Statement
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Octylphenol ethoxylate
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.