Learn More
Triton™ X-114, Pure, Fisher Chemical
Brand: Fisher Chemical T/3757/08
80.06 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Code : 27
Additional Details : CAS Number : 9002-93-1 Weight : 0.52750kg
Specifications
Clear | |
C14H22O·7·5(C2H4O) | |
500 mL |
9002-93-1 | |
537g/mol |