missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Trypsin-EDTA (0.05%), phenol red
Trypsin-EDTA (0.05%), phenol red
Brand: Gibco™ 25300104
Code : 50
Additional Details : Weight : 0.50000kg
Description
Gibco™ Trypsin-EDTA is made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas. Due to its digestive strength, trypsin is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation. The trypsin concentration required for dissociation varies with cell type and experimental requirements. See the complete range of Gibco™ trypsin solutions and recommended dissociation conditions.We offer a variety of trypsin formulations and animal origin-free TrypLE™ reagents that feature:
Quality Testing
Gibco™ Trypsin solutions are tested for pH, osmolality, sterility, and performance. In addition, prior to manufacturing, the raw materials are verified for e-beam irradiation and tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal, and viral contamination, as well as multiple activity assays, ash analysis, and moisture analysis.
Documented Traceability
We can provide detailed documentation to meet your regulatory needs. Gibco™ Trypsin information available includes lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency.
cGMP Manufacturing and Quality System
Gibco™ Trypsin is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Trypsin solution made in our Grand Island, New York facility (25300-120). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
ISO 13485 standards, FDA | |
Trypsin-EDTA, 10 x 500mL, 1 X Concentration, Liquid, 0.05% Dilution, With Phenol Red, EDTA Chelators, Porcine Pancreas Source, Trypsin Reagent Solution, FDA and ISO 13485 Standards, Used for Mammalian Cell Culture, –5 to –20°C Storage Tempe | |
1 X | |
Cell Culture Dissociation Reagent | |
Mammalian cell culture | |
For research use only |
–5 to –20°C | |
Mammalian Cells | |
Liquid | |
EDTA, Phenol Red | |
10 x 500 mL | |
Porcine pancreas |
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.