missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Trypsin-EDTA (0.05%), phenol red
Trypsin-EDTA (0.05%), phenol red
£238.00 - £500.00
Specifications
Certifications/Compliance | ISO 13485 standards, FDA |
---|---|
Content And Storage | –5 to –20°C |
Cell Type | Mammalian Cells |
Concentration | 1 X |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11500636
|
Gibco™
25300096 |
20 x 100mL |
£238.00
Pack of 20 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11510636
|
Gibco™
25300104 |
10 x 500 mL |
£500.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gibco™ Trypsin-EDTA is made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas. Due to its digestive strength, trypsin is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation. The trypsin concentration required for dissociation varies with cell type and experimental requirements. See the complete range of Gibco™ trypsin solutions and recommended dissociation conditions.We offer a variety of trypsin formulations and animal origin-free TrypLE™ reagents that feature:
Quality Testing
Gibco™ Trypsin solutions are tested for pH, osmolality, sterility, and performance. In addition, prior to manufacturing, the raw materials are verified for e-beam irradiation and tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal, and viral contamination, as well as multiple activity assays, ash analysis, and moisture analysis.
Documented Traceability
We can provide detailed documentation to meet your regulatory needs. Gibco™ Trypsin information available includes lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency.
cGMP Manufacturing and Quality System
Gibco™ Trypsin is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Trypsin solution made in our Grand Island, New York facility (25300-120). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
ISO 13485 standards, FDA | |
Mammalian Cells | |
Liquid | |
EDTA, Phenol Red | |
For research use only |
–5 to –20°C | |
1 X | |
Cell Culture Dissociation Reagent | |
Mammalian cell culture | |
Porcine pancreas |
Safety and Handling
missing translation for 'shelfLife' : 24 Months
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.