missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Trypsin, Porcine Pancreas
Typical use for this product is in removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture.
Specifications
For Use With (Application) | Release adherent cells from tissue culture plates for passaging |
---|---|
Sample Type | Porcine |
Content And Storage | 4°C |
Form | Powder |
Product Type | Trypsin |
Description
- CAS Number: 9002-07-7
- Molecular Weight: Approximately 23,000 or 22000 (for human)
- Composition: Trypsin is composed of two subunits, a-trypsin and b-trypsin. a-Trypsin is composed of two peptide chains and b-trypsin is composed of one chain.
- Activity: One BAEE unit will produce a DA253 of 0.001 per minute at pH 7.6 at 25°C using BAEE as substrate. Reaction volume = 3.2mL (1 cm light path).
One TAME unit hydrolyzes 1 umole of p-toluene-sulfonyl-L-arginine methyl ester (TAME) per minute at 25°C, pH 8.2, in the presence of 0.001 M calcium ion.
One USP trypsin unit is the activity causing a change in absorbance of 0.003 per minute under the conditions specified. - Activity Conversion:1 TAME unit = 19.2 USP or NF units = 57.5 BAEE Units
- Composition: Trypsin is composed of two subunits, a-trypsin and b-trypsin. a-Trypsin is composed of two peptide chains and b-trypsin is composed of one chain.
Specifications
Release adherent cells from tissue culture plates for passaging | |
4°C | |
Trypsin |
Porcine | |
Powder |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Trypsin 1:250, Virus Free
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : trypsin
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title