Learn More
BD Vacutainer™ Luer Adapters
Ideal for use with any female luer fitting. BD Vacutainer™ Luer Adapters are sterile devices with a multi-sampling valve that are designed for use with a catheter or hypodermic needle to collect blood using BD Vacutainer™ blood collection tubes.
Brand:  BD 367300
Product Code. 12967726
  • £788.00 / Pack of 1000
Explore more special offers
Description
Description
The BD Vacutainer™ system is a closed system optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process.
  • Provides sterile, secure and safe specimen sampling for fast, accurate and consistent results
  • Offers the security of a threaded, locking luer connection, replacing a luer-slip device
  • Latex-free and compatible with a female luer connection or needleless IV site designed for luer lock access
  • Used together with the BD Vacutainer™ single-use tube holder and evacuated blood collection tubes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Single-use
Vacutainer™ Single-use Tube Holders and Vacutainer™ Evacuated Blood Collection Tubes
Yes
Sterile
Blood collection from a catheter, needleless IV site, hypodermic needle or any female luer fitting; For safe and easy subculturing
With luer adapter
Tube Adapters and Holder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Vacutainer™ Luer Adapters > Color code: blue; Shelf Life: 1095 Days