missing translation for 'onlineSavingsMsg'
Learn More

Gosselin™ Tube Closures

Suitable for different usage of tubes and applications. Gosselin™ Tube Closures are offered in straight, winged, skirted, or screw cap styles and are manufactured from PE plastic.

Brand:  Gosselin™ BA110-23

89.09 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11592762

  • £103.00 / Pack of 2500
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • All closures vertically ribbed for easier grip
  • Protected with a polyethylene outer bag to prevent particle contamination
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EC N°1935/2004, ISO 11137
LDPE
11 mm Tube
Ribbed Closure Plug
White
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Corning Matrigel , Corning Biocoat , Corning CoolProduct , Corning Media Tech , Corning
    Corning Lab Products Offer
    Promo Code24073

    Expires2025-04-25

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gosselin™ Tube Closures > Winged Cap; Fits 11mm tube; White

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.