Learn More
Gosselin™ Tube Closures
Suitable for different usage of tubes and applications. Gosselin™ Tube Closures are offered in straight, winged, skirted, or screw cap styles and are manufactured from PE plastic.
Brand: Gosselin™ B185-12
Code : A0
Additional Details : Weight : 0.90000kg
Description
- All closures vertically ribbed for easier grip
- Protected with a polyethylene outer bag to prevent particle contamination
Specifications
Straight Plug | |
Red | |
Rack or Centrifuge |
LDPE | |
1000/Bag |