missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ MixMate Tube Holder

Designed for use with MixMate Vortex Mixer

Brand:  Eppendorf™ 5353040130

95.70 GBP valid until 2024-12-31
Use promo code "23292" to get your promotional price.



Code : 99

Additional Details : Weight : 0.18000kg

 View more versions of this product

Product Code. 10282963

  • £101.00 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Tube Holders for Eppendor MixMate™
  • The 3-in-1 mixer for plates, tubes and vortex mixing
  • High speed instrument guarantees complete mixing of your samples in seconds
  • Outstanding 2D Mix-Control provides optimized mixing with anti-spill technology
Specifications

Specifications

2 mL
Tube Holder
24 x 1.5mL/2mL micro test tubes
24
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers