Learn More
Eppendorf™ MixMate Tube Holder
Designed for use with MixMate Vortex Mixer
£101.00 - £104.42
Product Code | Brand | Tube Capacity | No. of Positions | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tube Capacity | No. of Positions | Price | Quantity & Availability | ||||
10130623
|
Eppendorf™
5353040113 |
0.2 mL | 96 |
£104.42
Each |
In Stock
Log in to see stock. |
||||
10691104
|
Eppendorf™
5353040121 |
0.5 mL | 24 |
£101.00
Each |
In Stock
Log in to see stock. |
||||
10282963
|
Eppendorf™
5353040130 |
2 mL | 24 |
£101.00
Each |
In Stock
Log in to see stock. |
||||
Description
- Tube Holders for Eppendor MixMate™
- The 3-in-1 mixer for plates, tubes and vortex mixing
- High speed instrument guarantees complete mixing of your samples in seconds
- Outstanding 2D Mix-Control provides optimized mixing with anti-spill technology
Specifications
Tube Holder |