Learn More
Cytiva Tube Holder
Tube Holder for Frac-900 or Frac-901
Brand:  Cytiva 19-8689-02
Code : GE
Additional Details : Weight : 0.41500kg
 View more versions of this product
Product Code. 12514437
  • £241.00 / 1 set
Estimated Shipment: 18-10-2024
to see stock.
Add to basket
Description
Description
The tube guide can be placed at different heights to accommodate different length of tubes.
Specifications
Specifications
Tube Holder
1 Pack
Tube holder and guide
95 Tubes