Learn More
Cytiva Tube Holder
Tube Holder for Frac-900 or Frac-901
£241.00 - £740.00
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
12554307
|
Cytiva
18-1124-68 |
Frac-900 |
£740.00
Pack of 2 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
12514437
|
Cytiva
19-8689-02 |
95 Tubes |
£241.00
1 set |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
Description
The tube guide can be placed at different heights to accommodate different length of tubes.Specifications
Tube Holder |