Learn More
Masterflex™ L/S™ Precision Tubing, PharMed™ BPT
Use this tubing for cell and tissue culture work and pharmaceutical applications. Masterflex™ L/S™ Precision Tubing, PharMed™ BPT resists acids, alkalies, oxidizing agents, and animal and vegetable oils. Tubing is heat-sealable, bondable and formable. Good for light-sensitive samples.
£135.00 - £650.00
Product Code | Brand | Tubing Size | Overall Length (English) | Diameter (English) Inner | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Overall Length (English) | Diameter (English) Inner | Material | Price | Quantity & Availability | |||
11852603
|
Masterflex™
06508-13 |
13 | 25 ft. | 1/16 in. | PharMed BPT |
£135.00
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||
11862603
|
Masterflex™
06508-14 |
14 | 25 ft. | 1/16 in. | PharMed BPT |
£234.00
7.50m |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||
11872603
|
Masterflex™
06508-16 |
16 | 25 ft. | 1/8 in. | PharMed BPT |
£595.00
7.50m |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||
11892603
|
Masterflex™
06508-17 |
17 | 25 ft. | 1/4 in. | PharMed BPT |
£297.00
Each |
Estimated Shipment: 13-06-2025
Log in to see stock. |
|||
11802613
|
Masterflex™
06508-18 |
18 | 25 ft. | 3/8 in. | PharMed BPT |
£224.00
Each |
Estimated Shipment: 04-08-2025
Log in to see stock. |
|||
11882603
|
Masterflex™
06508-25 |
25 | 25 ft. | 3/16 in. | PharMed BPT |
£650.00
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
Description
Use this tubing for cell and tissue culture work and pharmaceutical applications. PharMed™ BPT L/S™ Precision Pump Tubing resists acids, alkalies, oxidizing agents, and animal and vegetable oils. Tubing is heat-sealable, bondable, and formable. Also good for light-sensitive samples.
Masterflex™ L/S™ pump tubing is manufactured to extremely close tolerances that match our L/S™ pump heads, ensuring accurate, repeatable flow and long tubing life. Our pump tubing is factory-tested and optically inspected to provide the best performance from your peristaltic pump.
- Applications: Cell and tissue culture work and pharmaceutical uses. Also good for light-sensitive samples.
- Characteristics: Nontoxic; nonhemolytic; extremely low permeability; resists acids, alkalies, oxidizing agents, and animal and vegetable oils; heat-sealable, bondable, and formable. Opaque beige.
- Certifications: FDA compliant and meets USP Class VI and EP requirements. Tubing is manufactured according to GMP.
Specifications
25 ft. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.