Learn More
Tungsten gauze, gold plated, Thermo Scientific Chemicals
Description
Filtration, electrodesTungsten gauze, gold plated, wire is used in filtration. It also serves as an electrode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 150 x 150 mm |
Solubility Information | Soluble in mixture of nitric acid and hydrofluoric acid. Insoluble in water. |
Mesh Size | 50 Mesh |
Form | Wire Cloth |
Chemical Name or Material | Tungsten |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.