missing translation for 'onlineSavingsMsg'
Learn More

Tungsten sulfide, 99.8% (metals basis), Thermo Scientific Chemicals

£63.42 - £209.04

Chemical Identifiers

CAS 12138-09-9
Molecular Weight (g/mol) 247.96
MDL Number MFCD00011467
InChI Key ITRNXVSDJBHYNJ-UHFFFAOYSA-N
IUPAC Name disulfanylidenetungsten
SMILES S=[W]=S
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11311878
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
011829.18
50 g
£63.42
50g
Please to purchase this item. Need a web account? Register with us today!
11301878
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
011829.30
250 g
£209.04
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tungsten sulfide is used as a component of catalysts for hydrotreating of crude oil, hydrodesulfurization and hydrodenitrification reactions. It finds application as solid-state secondary lithium battery cathodes and other electrochemical devices. It is found that it forms inorganic nanotubes, which is better reinforcing agents than carbon nanotubes. Tungsten sulfide addition to epoxy resin improves adhesion, fracture toughness and strain energy release rate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12138-09-9
MFCD00011467
disulfanylidenetungsten
247.96
ITRNXVSDJBHYNJ-UHFFFAOYSA-N
S=[W]=S

Specifications

1250°C (decomposition)
Black
99.8% metal basis
MFCD00011467
ITRNXVSDJBHYNJ-UHFFFAOYSA-N
disulfanylidenetungsten
247.98
7.5
12138-09-9
Powder
S2W
Slightly soluble in water.
S=[W]=S
247.96
Odorless
Tungsten sulfide
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 235-243-3

missing translation for 'rtecsNumber' : YO7716000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only