missing translation for 'onlineSavingsMsg'
Learn More

Tungsten(VI) chloride, 99.9+%, (trace metal basis), Thermo Scientific Chemicals

£51.15 - £231.15

Chemical Identifiers

CAS 13283-01-7
Molecular Formula Cl6W
Molecular Weight (g/mol) 396.57
MDL Number MFCD00011463
InChI Key KPGXUAIFQMJJFB-UHFFFAOYSA-H
Synonym tungsten vi chloride, tungsten hexachloride, wolfram hexachloride, wcl6, tungsten chloride wcl6 , oc-6-11, tungsten chloride, oc-6-11, tungsten chloride 8ci, tungsten 6+ hexachloride, wolframhexachlorid, hexachloridotungsten
PubChem CID 83301
ChEBI CHEBI:37771
IUPAC Name hexachlorotungsten
SMILES Cl[W](Cl)(Cl)(Cl)(Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10667202
SDS View Documents
Thermo Scientific Acros
221430100
10 g Glass bottle
£51.15
10g
Please to purchase this item. Need a web account? Register with us today!
10772485
SDS View Documents
Thermo Scientific Acros
221431000
100 g Glass bottle
£231.15
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tungsten(VI) chloride, 99.9+%, (trace metal basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.