Learn More
Tungsten(VI) oxide, 99.8% (metals basis), Thermo Scientific Chemicals
£56.28 - £479.53
Chemical Identifiers
CAS | 1314-35-8 |
---|---|
Molecular Formula | O3W |
Molecular Weight (g/mol) | 231.837 |
MDL Number | MFCD00011466 |
InChI Key | ZNOKGRXACCSDPY-UHFFFAOYSA-N |
Synonym | tungsten trioxide, tungsten vi oxide, tungstic oxide, tungstic anhydride, tungsten oxide wo3, tungsten blue, tungstic acid anhydride, wolframic acid, anhydride, tungstentrioxide, acmc-20ajaaShow More |
PubChem CID | 14811 |
IUPAC Name | trioxotungsten |
SMILES | O=[W](=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11381868
|
Thermo Scientific Alfa Aesar
011828.22 |
100 g |
£56.28
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11391868
|
Thermo Scientific Alfa Aesar
011828.36 |
500 g |
£156.78
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11371868
|
Thermo Scientific Alfa Aesar
011828.A3 |
2 kg |
£479.53
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in tungsten and tungstate manufacturing which are used as X-ray screens and for fire proofing fabrics. It is used as a ceramic pigment. Nanowires of Tungsten (VI) oxide are capable of absorbing a higher percentage of the sun radiation since it absorbs blue light.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-35-8 | |
231.837 | |
ZNOKGRXACCSDPY-UHFFFAOYSA-N | |
14811 | |
O=[W](=O)=O |
O3W | |
MFCD00011466 | |
tungsten trioxide, tungsten vi oxide, tungstic oxide, tungstic anhydride, tungsten oxide wo3, tungsten blue, tungstic acid anhydride, wolframic acid, anhydride, tungstentrioxide, acmc-20ajaaShow More | |
trioxotungsten |
Specifications
1473°C | |
Powder | |
O3W | |
tungsten trioxide, tungsten vi oxide, tungstic oxide, tungstic anhydride, tungsten oxide wo3, tungsten blue, tungstic acid anhydride, wolframic acid, anhydride, tungstentrioxide, acmc-20ajaa | |
ZNOKGRXACCSDPY-UHFFFAOYSA-N | |
trioxotungsten | |
14811 | |
7.16 |
1314-35-8 | |
99.8% (metals basis) | |
MFCD00011466 | |
Tungsten (VI) oxide is soluble in hot alkali solutions. Insoluble in water and acids. Slightly soluble in hydrofluoric acid. | |
O=[W](=O)=O | |
231.837 | |
231.85 | |
Tungsten(VI) oxide |
Safety and Handling
missing translation for 'einecsNumber' : 215-231-4
missing translation for 'rtecsNumber' : YO7760000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.