Learn More
Abnova™ TurboTEV Protease
Tobacco etch virus (TEV) N1a protein catalytic fragment with GST and His tags expressed in Escherichia coli.
Brand: Abnova™ P5379.1mg
Additional Details : Weight : 0.02000kg
Specifications
2 mg/mL | |
Store at -20°C, Aliquot to avoid repeated freezing and thawing | |
1 mg |
Enzyme | |
Liquid | |
Protein |