Learn More
Thermo Scientific Chemicals Tween 20 washing buffer, 0.5% in PBS (10X)
Brand: Thermo Scientific Chemicals J60304.K2
Description
Tween 20 blocking buffer solution is used as a washing agent in immunoassays, such as Western blots and ELISAs. It helps to prevent non-specific antibody binding. The buffer is used for washes between each immunoreactions, to remove unbound immunologicals, and eventually for incubation solutions of immunoreagents (labeled antibodies) to reduce unspecific background.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Blocking Buffer | |
1 L | |
Liquid | |
0.5% in PBS (10X) |
RUO – Research Use Only