missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tween 20 washing buffer, 0.5% in PBS (10X)

£51.56 - £137.69

Specifications

Product Type Blocking Buffer
Form Liquid
Concentration 0.5% in PBS (10X)
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15443099
View Documents
Thermo Scientific Chemicals
J60304.K2
1 L
£75.38
1 L
Please to purchase this item. Need a web account? Register with us today!
15453099
View Documents
Thermo Scientific Chemicals
J60304.K3
2 L
£137.69
2 L
Please to purchase this item. Need a web account? Register with us today!
15433099
View Documents
Thermo Scientific Chemicals
J60304.AP
500 mL
£51.56
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tween 20 blocking buffer solution is used as a washing agent in immunoassays, such as Western blots and ELISAs. It helps to prevent non-specific antibody binding. The buffer is used for washes between each immunoreactions, to remove unbound immunologicals, and eventually for incubation solutions of immunoreagents (labeled antibodies) to reduce unspecific background.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Blocking Buffer
Liquid
0.5% in PBS (10X)
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only