missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tween™ 80 Surfact-Amps™ Detergent Solution

Thermo Scientific™ Surfact-Amps 80 Detergent Solution is highly-purified Tween-80 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.

Brand:  Thermo Scientific™ 28328

 View more versions of this product

Product Code. 11871455

  • £163.00 / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Tween-80 Detergent Surfact-Amps™ Solution is highly-purified Tween-80 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.

This is an easy-to-use 10% (w/v) solution of purified Tween-80 detergent for use in routine and high-demand protein research methods and molecular biology techniques. Tween-80 is a nonionic polyoxyethylene surfactant that is most frequently added to PBS or TBS wash buffers for ELISA, Western blotting and other immunoassay methods.

Surfact-Amps™ Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Highlights:

Tween 80 – a nonionic detergent for use in wash buffers

Accurate – precise 10% detergent solution in ultrapure water

Easy-to-use – solution is simple to dispense and dilute for use

Exceptionally pure – less than 1.0μeq/ml peroxides and carbonyls

Stable – packed in glass ampules under inert nitrogen gas

Detergent Properties:

  • Molecular Weight: 1310
  • Detergent Class: Nonionic
  • Aggregation Number: 60
  • Micelle Molecular Weight: 76,000
  • Critical Micelle Concentration (CMC): 0.012mM (0.0016%, w/v)
  • Cloud Point: Unknown
  • Dialyzable: No
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Detergent
Liquid
Detergent Solution
Store in a cool, dry, and well-ventilated area
Surfact-Amps™, Tween™
6 x 10 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.