Learn More
MP Biomedicals™ Tween™ 80, MP Biomedicals™
Tween™ 80 is widely used in a number of biochemical applications.
Brand: MP Biomedicals™ 02194725.1
Code : NEW
Additional Details : CAS Number : 9005-65-6 Weight : 0.10000kg
Description
Tweens™ are a series of nonionic surfactants derived from sorbitan esters.
- They are soluble or dispersible in water but differ widely in organic and oil solubilities.
- Used as oil-in-water emulsifiers in pharmaceuticals, cosmetics, cleaning compounds, etc.
- Polysorbate 20 is nonionic surfactants derived from sorbitan ester.
Specifications
Tween™ 80 | |
Polyoxyethylenesorbitan monooleate; POE (20) sorbitan monooleate; Polyethylene glycol sorbitan monooleate; Polysorbate 80 | |
Liquid | |
100 mL |
9005-65-6 | |
Store at Room Temperature | |
Selective protein extraction; Isolation of nuclei from mammalian cell lines |
Safety and Handling
- Tween 80, cell culture reagent
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : tween 80