Learn More
Labplas Twirl'EM™ Large Format Sampling Bags with Tabs
Polyethylene sampling bags with large openings
Brand:  Labplas EDL41218
 View more versions of this product
Product Code. 16336378
  • £171.00 / Pack of 250
Estimated Shipment: 24-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Polyethylene sampling bags with large openings
  • Large sized
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
Specifications
Specifications
Regular Tabs
2 Flat Wires with Tabs
Sterile
18 x 12 in. (457 x 305mm)
457 mm
305 mm
6.75 L
Polyethylene
4 mil
18 in.
12 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Labplas Twirl'EM™ Large Format Sampling Bags with Tabs > Regular tabs; Clear; 2 flat wires; 305 x 457mm; 6.75L volume