Learn More
Labplas Twirl'EM™ Large Format Sampling Bags with Tabs
Polyethylene sampling bags with large openings
£171.00
Specifications
Description Regular Tabs
Material Polyethylene
Sterility Sterile
Thickness (English) 4 mil
View More Specs
Products 1
Product Code Brand Capacity (Metric) Closure Type Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Type Price Quantity & Availability  
16336378
View Documents
Labplas
EDL41218
6.75 L 2 Flat Wires with Tabs
£171.00
Pack of 250
Estimated Shipment: 01-07-2025
to see stock.
Add to basket
Description
Description
Polyethylene sampling bags with large openings
  • Large sized
  • Contaminant-free
  • Virgin polyethylene
  • Large bag mouth opening
  • Easy to use pull tabs
Specifications
Specifications
Regular Tabs
Polyethylene
Sterile
4 mil
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Labplas Twirl'EM™ Large Format Sampling Bags with Tabs