missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Two-Stage Rotary Vane Pump FRVP Series

Rugged, must-have pumps for deep vacuum operation give high reliability and low maintenance operation in a wide range of lab and industrial applications

Brand:  Fisherbrand™

Additional Details : Weight : 38.70000kg

 View more versions of this product

Product Code. 16254611

  • £4054.50 / Each
Estimated Shipment: 04-10-2024
to see stock.
Add to basket

Description

Description

  • Designed for longevity and reliability
  • Constructed from high grade materials and corrosion-resistant coating on internal surfaces
  • Built for cleaner working environment
  • Reduced oil mist emissions
  • Cool running temperatures
  • Internal pre-filters and oil mist filters included (except FRVP28)
  • Quick and simple serviceability with mistake-proof poka-yoke servicing design
  • Use premium oil for quick and simple oil changes with extended oil-change intervals
  • Oil levels easily monitored through large viewing port
Specifications

Specifications

Connection flanges DN 25 KF, Motor protection level IP 54, Accessories: 3 L premium oil, Exhaust oil mist filter, 20 mm hose nozzle adapter, Centering and clamping rings
230 V, 50/60 Hz
FRVP16
700 W
280 L/min. (at 50 Hz)
Two-stage Rotary Vane Vacuum Pump
Chemistry: Schlenk lines, glove boxes, Life sciences: freeze dryers, vacuum concentrators, vacuum ovens (<0.1 mbar), Industrial: vacuum furnaces, isolation vacuums, Analytical: as replacement backing pumps for mass spectrometers, General R&D: for evacuation of vacuum systems and chambers down to fine vacuum levels (10-3mbar)
230 V
1450 rpm
569 x 206 x 289 mm (without handle and accessories)
< 50dB(A)
37 kg
Deep vacuum operation
2400 mL
Without gas ballast partial: 3 x 10-4mbar, Without gas ballast total: 3 x 10-3mbar, With gas ballast total: 6 x 10-3mbar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers