Learn More
Thermo Scientific™ TX-200 Rotor Tube Adapters
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Brand:  Thermo Scientific™ 75003809
 View more versions of this product
Product Code. 10799305
  • £244.96 / Pack of 4
Explore more special offers
Description
Description
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Adapters hold a variety of tubes including round-bottom, conical, urine, blood collection and microtubes.
  • Maximum tube capacity: 4 × 100mL
  • Maximum O.D. × L: 48 × 122mm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
17 mm
121 mm
15mL Blood Collection tube