Learn More
Thermo Scientific™ TX-200 Rotor Tube Adapters
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
£231.39 - £249.19
Products 15
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10194494
View Documents
Thermo Scientific™
75003771
15mL Conical tube
£231.39
Pack of 4
10596214
View Documents
Thermo Scientific™
75003785
5/7mL Blood Collection tube
£231.39
Pack of 4
10596404
View Documents
Thermo Scientific™
75003801
100mL Round-bottom tube
£245.80
Pack of 4
10338204
View Documents
Thermo Scientific™
75003802
50mL DIN Round-bottom tube
£245.80
Pack of 4
10082534
View Documents
Thermo Scientific™
75003803
50mL Conical tube
£231.39
Pack of 4
10216854
View Documents
Thermo Scientific™
75003804
25mL Universal tube
£244.96
Pack of 4
10297374
View Documents
Thermo Scientific™
75003805
25mL DIN Round-/Flat-bottom tube
£245.80
Pack of 4
10459834
View Documents
Thermo Scientific™
75003806
20mL Round-bottom tube This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10124364
View Documents
Thermo Scientific™
75003807
14mL Conical Urine tube
£246.65
Pack of 4
10318064
missing translation for 'viewDocuments'
Thermo Scientific™
75003808
10mL Blood Collection or 15mL Corex /Kimble tube
£231.39
Pack of 4
10799305
missing translation for 'viewDocuments'
Thermo Scientific™
75003809
15mL Blood Collection tube
£244.96
Pack of 4
10677565
missing translation for 'viewDocuments'
Thermo Scientific™
75003810
5/7mL Round-bottom tube
£245.80
Pack of 4
10267044
missing translation for 'viewDocuments'
Thermo Scientific™
75003811
12 x 124mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10431565
missing translation for 'viewDocuments'
Thermo Scientific™
75003812
1.5/2.0mL Conical Microtube
£249.19
Pack of 4
10738545
missing translation for 'viewDocuments'
Thermo Scientific™
75003815
50mL Round-bottom tube This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Adapters hold a variety of tubes including round-bottom, conical, urine, blood collection and microtubes.
  • Maximum tube capacity: 4 × 100mL
  • Maximum O.D. × L: 48 × 122mm