missing translation for 'onlineSavingsMsg'
Learn More

Ismatec™ Tygon™ MHSL 2001 2-stop Tubing

Transparent, plasticizer-free 2-stop Tygon™ tubing with superior pump-life. Ismatec™ Tygon™ MHSL 2001 2-stop Tubing is especially designed for MEK and other aggressive solvents.

Brand:  Ismatec™ SC0824

Code : NEW

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10114982

  • £117.00 / Pack of 6
Estimated Shipment: 29-10-2024
to see stock.
Add to basket

Description

Description

  • Plasticizer and oil-free.
  • Smooth inner-bore.
  • Low sorption maintains fluid and tube integrity.
  • Does not impart anything into the pumping medium.
  • No release of hazardous materials when properly incinerated.
  • Applications
    • Acids: Excellent
    • Alkaline solutions: Excellent
    • Solvents: Good/Excellent
  • Complies with: FDA.
  • Sterilization
    • Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C.
    • Gas sterilization with ethylene oxide.
    • Sterilization with radiation up to 2.5 mrad.
Specifications

Specifications

2-stop Tubing
Color-code: Purple-White
2.79 mm
4.49 mm
0.85 mm
For MEK and other aggressive solvents. CA cassettes
6 pcs./Pk.
2.79mm I.D., 4.49mm O.D.
Transparent
FDA
Autoclavable
Tygon MHSL 2001
38.1 cm
peristaltic pump
-73°C
Acids, Alkalies
CO2 1140, O2 76, N2 190
57°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers