missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ismatec™ Tygon™ MHSL 2001 2-stop Tubing
Transparent, plasticizer-free 2-stop Tygon™ tubing with superior pump-life. Ismatec™ Tygon™ MHSL 2001 2-stop Tubing is especially designed for MEK and other aggressive solvents.
£87.55 - £117.00
Specifications
Product Type | 2-stop Tubing |
---|---|
Certifications/Compliance | FDA |
Autoclavable | Autoclavable |
Material | Tygon MHSL 2001 |
Overall Length (Metric) | 38.1 cm |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
10114982
|
Ismatec™
SC0824 |
2.79mm I.D., 4.49mm O.D. |
£117.00
Pack of 6 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10454192
|
Ismatec™
SC0822 |
2.06mm I.D., 3.76mm O.D. |
£117.00
Pack of 6 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10423812
|
Ismatec™
SC0816 |
0.64mm I.D., 2.44mm O.D. |
£87.55
Pack of 6 |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
10338092
|
Ismatec™
SC0820 |
1.52mm I.D., 3.22mm O.D. |
£117.00
Pack of 6 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10504223
|
Ismatec™
SC0818 |
1.02mm I.D., 2.72mm O.D. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Plasticizer and oil-free.
- Smooth inner-bore.
- Low sorption maintains fluid and tube integrity.
- Does not impart anything into the pumping medium.
- No release of hazardous materials when properly incinerated.
- Applications
- Acids: Excellent
- Alkaline solutions: Excellent
- Solvents: Good/Excellent
- Complies with: FDA.
- Sterilization
- Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C.
- Gas sterilization with ethylene oxide.
- Sterilization with radiation up to 2.5 mrad.
Specifications
2-stop Tubing | |
Autoclavable | |
38.1 cm | |
For MEK and other aggressive solvents. CA cassettes | |
6 pcs./Pk. | |
CO2 1140, O2 76, N2 190 | |
57°C |
FDA | |
Tygon MHSL 2001 | |
peristaltic pump | |
-73°C | |
Acids, Alkalies | |
Transparent |