Learn More
Sartorius™ Type 144 Hydrosart™ Regenerated Cellulose (RC) Ultrafiltration Membranes
Type 144 Hydrosart™ Regenerated Cellulose (RC) Ultrafiltration Membranes are highly hydrophilic and have extended chemical compatibility. Easy to clean and autoclavable, they are ideal for protein recovery from dilute samples and work under aseptic conditions.
Brand: Sartorius™ 14429--44------D
156.18 GBP valid until 2024-12-31
Use promo code "23270" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Type: Ultrafiltration Membrane
- Material: Hydrosart™ Regenerated Cellulose (RC)
- Grade: Type 144
- Shape: Disc
- Flow Rate: 0.08 mL/min./cm2
- Thickness: 180 μm
- MWCO: 5 kDa
- For Use With (Application): Concentration, Protein Removal, Diafiltration, Desalting, Buffer Exchange, and Electro-Ultrafiltration (in BioPharma, Research, and Environmental Fields)
Specifications
Autoclavable | |
Disc | |
Hydrosart Regenerated Cellulose | |
Non-sterile | |
Concentration, Protein Removal, Diafiltration, Desalting, Buffer Exchange, and Electro-Ultrafiltration | |
Ultra-filtration Membrane Discs | |
10 Pk. | |
7.1 mil | |
Ultrafiltration Membrane Discs | |
44 mm |
Ultra-filtration Membrane Disc | |
Hydrosart | |
5 kDa | |
0.08 mL/min./cm2 | |
Type 144 | |
10 Pk. | |
Disc | |
180 μm | |
44 mm |