missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Type 144 Hydrosart™ Regenerated Cellulose (RC) Ultrafiltration Membranes
Type 144 Hydrosart™ Regenerated Cellulose (RC) Ultrafiltration Membranes are highly hydrophilic and have extended chemical compatibility. Easy to clean and autoclavable, they are ideal for protein recovery from dilute samples and work under aseptic conditions.
£182.00 - £284.00
Specifications
Autoclavable | Autoclavable |
---|---|
Filter Type | Ultra-filtration Membrane Disc |
Format | Disc |
Material | Hydrosart |
Material (Membrane) | Hydrosart Regenerated Cellulose |
Product Code | Brand | Diameter (Metric) Membrane | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Membrane | Price | Quantity & Availability | |||||
10205901
|
Sartorius™
14439--63------D |
63 mm |
£284.00
Pack of 10 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
11750925
|
Sartorius™
14429--44------D |
44 mm |
£182.00
Pack of 10 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
10032431
|
Sartorius™
14439--47------D |
47 mm |
£182.00
Pack of 10 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
Description
- Type: Ultrafiltration Membrane
- Material: Hydrosart™ Regenerated Cellulose (RC)
- Grade: Type 144
- Shape: Disc
- Flow Rate: 0.08 mL/min./cm2
- Thickness: 180 μm
- MWCO: 5 kDa
- For Use With (Application): Concentration, Protein Removal, Diafiltration, Desalting, Buffer Exchange, and Electro-Ultrafiltration (in BioPharma, Research, and Environmental Fields)
Specifications
Autoclavable | |
Disc | |
Hydrosart Regenerated Cellulose | |
Non-sterile | |
Concentration, Protein Removal, Diafiltration, Desalting, Buffer Exchange, and Electro-Ultrafiltration | |
Ultra-filtration Membrane Discs | |
10 Pk. | |
7.1 mil | |
Ultrafiltration Membrane Discs |
Ultra-filtration Membrane Disc | |
Hydrosart | |
5 kDa | |
0.08 mL/min./cm2 | |
Type 144 | |
10 Pk. | |
Disc | |
180 μm |