missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 30 Safety Storage Cabinet Q-DISPLAY-30

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3278106033622

Additional Details : Weight : 229.41901kg

 View more versions of this product

Product Code. 15937496

  • £2698.23 / Each
Estimated Shipment: 10-01-2025
to see stock.

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: triple hinged door, cabinet body made of melamine resin and flame retardant special plates, safety elements assembled outside the  storage compartment for increased protection against corrosion, doors with metal front as well as scratch-and impact-resistant surface, easy to clean
  • All containers at a glance: door with large glass cutout allows a quick and easy overview of the stored containers – even with closed doors
  • Easy handling with comfort: door stops in any position, self-closing in the event of fire
  • No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
  • Easy transport: integrated transport base with removable, optional base cover or adjusting aids with base cover
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications

Specifications

1626 mm
Safety storage cabinet
750 mm
Powder-coated textured
864 mm
620 x 864 x 1947 mm
519 x 750 x 1626 mm
Standard
1947 mm
Q30.195.086.WDFW
90 mm
Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
3 x shelf, 1 x perforated insert, 1 x 345 mm bottom collecting sump (sheet steel powder-coated)
Safe and approved storage of flammable hazardous substances in working areas
519 mm
Steel
Entry height transport base
237.50 kg
Anthracite Gray, Warning Yellow
620 mm
Fixed Window, Left-hinged
3
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers