missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 30 Safety Storage Cabinet Q-DISPLAY-30
Safe and approved storage of flammable hazardous substances in working areas
£2698.23 - £3783.14
Specifications
Certifications/Compliance | Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 620 mm |
Depth (Metric) Interior | 519 mm |
Dimensions (D x W x H) | 620 x 864 x 1947 mm |
Dimensions (D x W x H) Interior | 519 x 750 x 1626 mm |
Alert:
For indoor use only
Product Code | Brand | Configuration | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Color | Price | Quantity & Availability | ||||
15937496
|
asecos™
3278106033622 |
3 x shelf, 1 x perforated insert, 1 x 345 mm bottom collecting sump (sheet steel powder-coated) | Anthracite Gray, Warning Yellow |
£2698.23
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15977496
|
asecos™
3278106033627 |
4 x drawer (sheet steel powder-coated) | Anthracite Gray, Warning Yellow |
£3340.06
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15997496
|
asecos™
3278106033629 |
6 x drawer (sheet steel powder-coated) | Anthracite Gray, Warning Yellow |
£3783.14
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15907526
|
asecos™
3278106133622 |
3 x shelf, 1 x perforated insert, 1 x 345 mm bottom collecting sump (sheet steel powder-coated) | Anthracite Gray, Light Gray |
£2698.23
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15947526
|
asecos™
3278106133627 |
4 x drawer (sheet steel powder-coated) | Anthracite Gray, Light Gray |
£3340.06
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15967526
|
asecos™
3278106133629 |
6 x drawer (sheet steel powder-coated) | Anthracite Gray, Light Gray |
£3783.14
Each |
Estimated Shipment: 10-01-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Robust construction and longevity: triple hinged door, cabinet body made of melamine resin and flame retardant special plates, safety elements assembled outside the storage compartment for increased protection against corrosion, doors with metal front as well as scratch-and impact-resistant surface, easy to clean
- All containers at a glance: door with large glass cutout allows a quick and easy overview of the stored containers – even with closed doors
- Easy handling with comfort: door stops in any position, self-closing in the event of fire
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy transport: integrated transport base with removable, optional base cover or adjusting aids with base cover
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
519 mm | |
519 x 750 x 1626 mm | |
Steel | |
Fixed Window, Left-hinged | |
1947 mm | |
Entry height transport base | |
1 | |
864 mm |
620 mm | |
620 x 864 x 1947 mm | |
90 mm | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1626 mm | |
Q30.195.086.WDFW | |
Powder-coated textured | |
750 mm |