missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 30 Safety Storage Cabinet Q-DISPLAY-30
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 3278106133627
2654.35 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 255.51900kg
Alert:
For indoor use only
Description
- Robust construction and longevity: triple hinged door, cabinet body made of melamine resin and flame retardant special plates, safety elements assembled outside the storage compartment for increased protection against corrosion, doors with metal front as well as scratch-and impact-resistant surface, easy to clean
- All containers at a glance: door with large glass cutout allows a quick and easy overview of the stored containers – even with closed doors
- Easy handling with comfort: door stops in any position, self-closing in the event of fire
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy transport: integrated transport base with removable, optional base cover or adjusting aids with base cover
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
1626 mm | |
Safety storage cabinet | |
750 mm | |
Powder-coated textured | |
864 mm | |
620 x 864 x 1947 mm | |
519 x 750 x 1626 mm | |
620 mm | |
Fixed Window, Left-hinged | |
90 mm | |
Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
4 x drawer (sheet steel powder-coated) | |
Safe and approved storage of flammable hazardous substances in working areas | |
519 mm | |
Steel | |
Entry height transport base | |
266.50 kg | |
Anthracite Gray, Light Gray | |
1947 mm | |
Q30.195.086.WDFW | |
1 |