missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Combi Safety Storage Cabinet
Safety storage cabinet
Brand: asecos™ 30335-001-30755
3098.16 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 551.50000kg
Alert:
For indoor use only
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Two in one: 2 compartments (vertical seperated) allow the storage of flammable liquids and aggressive chemicals in one cabinet
- No unauthorized use: doors lockable with profile cylinder (integration in an existing locking system possible) /cylinder lock
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection to a technical exhaust system
- Fire resistance 90 minutes
- type tested in accordance with EN14470-1
- DIN EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
Combi safety storage cabinet K-PHOENIX-90 model K90.196.120.MC.FWAS light gray (RAL 7035), interior equipment with 1 x storage box (sheet steel powder coated), 2 x shelf (stainless steel), 1 x perforated insert, 1 x bottom collecting sump (stainless steel), 4 x pull-out shelf (panel melamine resin-coated/PP) | |
Plain | |
Cabinet and interior equipment, Door-open Arrest System | |
24.29 in. | |
617 mm | |
617 mm | |
Combi safety storage cabinet | |
47.13 in. | |
Light Gray | |
1969 mm | |
K90.196.120.MC.FWAS | |
1742 mm (Left), 862 mm (Right) | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
77.52 in. | |
Gypsum | |
617 x 1197 x 1969 mm | |
522 x 450 x 1742 mm (left), 542 x 491 x 862 mm (right top and bottom) | |
30 kg, 75 kg, 25 kg | |
2, 4 | |
Safe and approved storage of aggressive and flammable hazardous substances in working areas | |
1197 mm | |
Pull-out Shelves | |
Wing | |
1197 mm | |
3 | |
443 mm |