missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Combi Safety Storage Cabinet
Safety storage cabinet
£4091.60
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) | 617 mm |
Depth (Metric) Exterior | 617 mm |
Color | Light Gray |
Depth (English) Exterior | 24.29 in. |
Alert:
For indoor use only
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
17829373
|
asecos™
30335-001-30755 |
K90.196.120.MC.FWAS |
£4091.60
Each |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Two in one: 2 compartments (vertical seperated) allow the storage of flammable liquids and aggressive chemicals in one cabinet
- No unauthorized use: doors lockable with profile cylinder (integration in an existing locking system possible) /cylinder lock
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection to a technical exhaust system
- Fire resistance 90 minutes
- type tested in accordance with EN14470-1
- DIN EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
617 mm | |
24.29 in. | |
522 x 450 x 1742 mm (left), 542 x 491 x 862 mm (right top and bottom) | |
1742 mm (Left), 862 mm (Right) | |
Combi safety storage cabinet | |
Wing | |
1969 mm | |
30 kg, 75 kg, 25 kg | |
Pull-out Shelves | |
1197 mm | |
443 mm |
617 mm | |
Light Gray | |
617 x 1197 x 1969 mm | |
77.52 in. | |
Gypsum | |
47.13 in. | |
Safe and approved storage of aggressive and flammable hazardous substances in working areas | |
Cabinet and interior equipment, Door-open Arrest System | |
3 | |
Plain | |
1197 mm |