Learn More
asecos Type 90 Combi Safety Storage Cabinet K-PHOENIX-90
Safe and approved storage of aggressive and flammable hazardous substances in working areas
Brand: asecos 30333-001-34257
3094.72 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 490.57919kg
Alert:
For indoor use only
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Two in one: 2 compartments (vertical separated) allow the storage of flammable liquids and aggressive chemicals in one cabinet
- User-friendly and safe (folding door compartment): door open arrest system (standard), convenient and safe removal or storage of containers
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)/cylinder lock
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Certificates
- Fire resistance: 90 minutes
- Type tested in accordance with EN 14470-1
- DIN EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
Combi Safety Storage Cabinet | |
77.52 in. | |
Plain | |
1197 mm | |
617 x 1197 x 1969 mm | |
24.29 in. | |
Light Gray | |
617 mm | |
1146 lb. | |
Wing, Folding | |
3 (left)/4 (right) | |
2 |
Safe and approved storage of aggressive and flammable hazardous substances in working areas | |
47.13 in. | |
Gypsum | |
(3) tray shelves, (1) bottom collecting sump (sheet steel powder-coated), (4) pull-out shelves (panel/PP) | |
520 kg | |
Right top: 542 x 491 x 862 mm; Right bottom: 542 x 491 x 862 mm; Left: 524 x 450 x 1742 mm | |
Standard/pull-out | |
1969 mm | |
150 kg (tray shelf)/25 kg (pull-out shelf) | |
K90.196.120.MF.FWAS | |
Right: 395 mm (all); Left: 500/350/315/500 mm | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |