missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos Type 90 Combi Safety Storage Cabinet K-PHOENIX-90
Safe and approved storage of aggressive and flammable hazardous substances in working areas
£3911.40 - £4997.90
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 617 mm |
Color | Light Gray |
Depth (English) Exterior | 24.29 in. |
Dimensions (D x W x H) | 617 x 1197 x 1969 mm |
Alert:
For indoor use only
Product Code | Brand | No. of Shelves | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Shelves | Includes | Price | Quantity & Availability | ||||
15914316
|
asecos
3033500133841 |
2 (left)/4 (right) | (1) storage box, (2) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated), (4) pull-out shelves (panel melamine resin-coated/PP) |
£4150.43
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15964286
|
asecos
30333-001-34257 |
3 (left)/4 (right) | (3) tray shelves, (1) bottom collecting sump (sheet steel powder-coated), (4) pull-out shelves (panel/PP) |
£4330.10
Each |
Estimated Shipment: 19-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15690391
|
asecos
3033300133838 |
3 (left)/4 (right) | (3) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated), (4) pull-out shelves (panel melamine resin-coated/PP) |
£3911.40
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15944286
|
asecos
3033300133840 |
6 (left)/ 6 (right) | (6) drawers (stainless steel), (6) pull-out shelves (panel melamine resin-coated/PP) |
£4997.90
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Two in one: 2 compartments (vertical separated) allow the storage of flammable liquids and aggressive chemicals in one cabinet
- User-friendly and safe (folding door compartment): door open arrest system (standard), convenient and safe removal or storage of containers
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)/cylinder lock
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Certificates
- Fire resistance: 90 minutes
- Type tested in accordance with EN 14470-1
- DIN EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
Light Gray | |
617 x 1197 x 1969 mm | |
77.52 in. | |
Combi Safety Storage Cabinet | |
Wing, Folding | |
1969 mm | |
Plain |
617 mm | |
24.29 in. | |
Right top: 542 x 491 x 862 mm; Right bottom: 542 x 491 x 862 mm; Left: 524 x 450 x 1742 mm | |
Gypsum | |
47.13 in. | |
Safe and approved storage of aggressive and flammable hazardous substances in working areas | |
2 | |
1197 mm |