missing translation for 'onlineSavingsMsg'
Learn More

asecos Type 90 Combi Safety Storage Cabinet K-PHOENIX-90

Safe and approved storage of aggressive and flammable hazardous substances in working areas

Brand:  asecos 3033300133838

3664.20 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Code : NEW

Additional Details : Weight : 515.00000kg

 View more versions of this product

Product Code. 15690391

  • £3911.40 / Each
Estimated Shipment: 03-12-2024
to see stock.

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • Two in one: 2 compartments (vertical separated) allow the storage of flammable liquids and aggressive chemicals in one cabinet
  • User-friendly and safe (folding door compartment): door open arrest system (standard), convenient and safe removal or storage of containers
  • No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)/cylinder lock
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system

Certificates

  • Fire resistance: 90 minutes
  • Type tested in accordance with EN 14470-1
  • DIN EN 16121/16122 conformity
  • GS certified
  • CE compliant
Specifications

Specifications

Combi Safety Storage Cabinet
77.52 in.
Plain
1197 mm
617 x 1197 x 1969 mm
24.29 in.
Light Gray
617 mm
1155 lb.
Wing, Folding
3 (left)/4 (right)
2
Safe and approved storage of aggressive and flammable hazardous substances in working areas
47.13 in.
Gypsum
(3) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated), (4) pull-out shelves (panel melamine resin-coated/PP)
524 kg
Right top: 542 x 491 x 862 mm; Right bottom: 542 x 491 x 862 mm; Left: 524 x 450 x 1742 mm
Standard/pull-out
1969 mm
30 kg (standard shelf)/75 kg (grid shelf)/25 kg (pull-out shelf)
K90.196.120.MF.FWAS
Right: 395 mm (all); Left: 380/410/380/435 mm
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers